N-(Hydroxymethyl)phthalimide, Sigma-Aldrich®

Supplier: Merck

Synonyms: 2-(Hydroxymethyl)isoindoline-1,3-dione

M-Clarity™ quality level = MQ200

8.14422.0100EA 57.3 EUR
8.14422.9999 8.14422.0100
N-(Hydroxymethyl)phthalimide, Sigma-Aldrich®
N-(Hydroxymethyl)phthalimide
Formula: C₉H₇NO₃
MW: 177.16 g/mol
Boiling Pt: 415 °C (1013 hPa)
Melting Pt: 149…150 °C
MDL Number: MFCD00005899
CAS Number: 118-29-6
EINECS: 204-241-4

Order Now

Specification Test Results

Solubility (20 °C) slightly soluble
Melting Point 149 - 150 °C
Molar Mass 177.16 g/mol
Hill Formula C9H7NO3

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR