EDTA tripotassium salt

Supplier: Molekula

Synonyms: Edta-3k,Ethylenediaminetetra-acetic acid tripotassium salt,Tripotassium hydrogen ethylenediaminetetraacetate,Ethylenedinitrilotetra-acetic acid tripotassium salt,Edetic acid tripotassium salt

52369963-100G
MOLE52369963-100GEA 0 EUR
MOLE52369963-100G
EDTA tripotassium salt
EDTA tripotassium salt
Formula: C₁₀H₁₃K₃N₂O₈
MW: 406.52 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00013304
CAS Number: 17572-97-3
EINECS: 241-543-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR