4,4'-Dichlorobenzhydrol

Supplier: Molekula

MOLE56493615-100GEA 0 EUR
MOLE56493615-100G
4,4'-Dichlorobenzhydrol
4,4'-Dichlorobenzhydrol
Formula: C₁₃H₁₀Cl₂O
MW: 253.13 g/mol
MDL Number: MFCD00000629
CAS Number: 90-97-1
EINECS: 202-029-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR