(Ethyl)triphenylphosphonium iodide

Supplier: Molekula

Synonyms: Ethyltriphenylphosphonium iodide

11076392-100G
MOLE11076392-100GEA 0 EUR
MOLE11076392-100G
(Ethyl)triphenylphosphonium iodide
(Ethyl)triphenylphosphonium iodide
Formula: C₂₀H₂₀IP
MW: 418.26 g/mol
Melting Pt: 163…166 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00040352
CAS Number: 4736-60-1
EINECS: 225-245-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR