2-Deoxy-L-ribose

Supplier: Molekula

Synonyms: 2-Deoxy-L-erythropentose,2-Deoxy-L-arabinose,2-Deoxy-L(+)-ribose

M77942583
MOLEM77942583EA 0 EUR
MOLEM77942583
2-Deoxy-L-ribose
2-Deoxy-L(+)-ribose
Formula: C₅H₁₀O₄
MW: 134.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00132941
CAS Number: 18546-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR