Uracil

Supplier: Molekula

Synonyms: Pyrimidine-2,4(1H,3H)-dione

31200749-100G
MOLE31200749-100GEA 0 EUR
MOLE31200749-100G
Uracil
Uracil
Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Boiling Pt: 379 °C (1013 hPa)
Melting Pt: 194 °C
Density: 0.99744 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006016
CAS Number: 66-22-8
EINECS: 200-621-9
Merck Index: 12,09985

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR