Diethylene glycol monoethyl ether ≥98%

Warning

Synonyms: 2-(2-Ethoxyethoxy)ethanol,Di(ethylene glycol) ethyl ether,Diethylene Glycol Monoethyl Ether

A14670.0C A14670.0E A14670.36
A14670.0CEA 158 EUR
A14670.0C A14670.0E A14670.36
Diethylene glycol monoethyl ether ≥98%
Diethylene glycol monoethyl ether
Formula: C₆H₁₄O₃
MW: 134.18 g/mol
Boiling Pt: 201…202 °C
Melting Pt: –80 °C
Density: 0.989
Flash Pt: 96 °C (204 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002872
CAS Number: 111-90-0
EINECS: 203-919-7
REACH: 01-2119475105-42
Merck Index: 13,01809

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR