Citral

Supplier: Roth Carl
Warning

Synonyms: 3,7-Dimethylocta-2,6-dienal

5292.1
ROTH5292.1EA 139 EUR
ROTH5292.1
Citral
Citral
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 229 °C (1013 hPa)
Melting Pt: < –10 °C
Density: 0,89 g/cm³ (20 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00006997
CAS Number: 5392-40-5
EINECS: 226-394-6
Merck Index: 13,02346

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR