Isopentyl hexanoate

Supplier: Merck

Synonyms: Isopentyl hexanoate,Isoamyl caproate,Isoamyl hexanoate,Hexanoic acid Isopentyl ester,Hexanoic acid isoamyl ester,Isopentyl caproate,Caproic acid isoamyl ester,Caproic acid isopentyl ester,3-Methylbutyl hexanoate,Hexanoic acid isopentyl ester

M-Clarity™ quality level = MQ200

8.14994.0250
8.14994.0250EA 96.7 EUR
8.14994.0250
Isopentyl hexanoate
Isopentyl hexanoate
Formula: C₁₁H₂₂O₂
MW: 186.29 g/mol
Boiling Pt: 226 °C (1013 hPa)
Density: 0.861 g/cm³ (20 °C)
MDL Number: MFCD00027280
CAS Number: 2198-61-0
EINECS: 218-600-8

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Molar Mass 186.3 g/mol
Boiling Point 222 °C (1013 hPa)
Flash Point 85 °C
Density 0.861 g/cm3 (20 °C)
Synonyms Methylbutyl caproate, Caproic acid methylbutyl ester, Hexanoic acid methylbutyl ester
Hill Formula C11H22O2

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR