(R)-(+)-1,2-Epoxypropane, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: (R)-(+)-Propene oxide,(R)-(+)-Methyloxirane,(R)-(+)-1,2-Propylene oxide,(R)-(+)-Propylene oxide

M-Clarity™ quality level = MQ200

8.18567.0001
8.18567.0001EA 206 EUR
8.18567.0001
(R)-(+)-1,2-Epoxypropane, Sigma-Aldrich®
(R)-(+)-1,2-Epoxypropane
Formula: C₃H₆O
MW: 58.08 g/mol
Density: 0.829 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00066211
CAS Number: 15448-47-2
EINECS: 200-879-2
UN: 1280
ADR: 3,I

Order Now

Specification Test Results

Solubility 405 g/l (20 °C)
Melting Point -112 °C
Molar Mass 58.07 g/mol
Boiling Point 34 - 35 °C (1013 hPa)
Vapor Pressure 588 hPa (20 °C)
Flash Point -37 °C
Refractive Index 1.366 (20 °C, 589 nm)
Explosion Limit 1.9 - 37 %(V)
Density 0.829 g/cm3 (20 °C)
Ignition Point 430 °C DIN 51794

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR