Hydrochloric acid 3 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Supplier: MERCK PRODUCTION CHEMICALS
Warning
4.80680.9025
4.80680.9025EA 0 EUR
4.80680.9025
Hydrochloric acid 3 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Density: 1.06 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II