Bromophenol blue 0.1% in aqueous solution
Supplier: Bernd Kraft
Synonyms: Bromphenol Blue Sultone Form,BPB,3,3',5,5'-Tetrabromophenolsulphonphthalein,Tetrabromophenol-sulphonephthalein
03012.1000 03012.2000 03012.3000
KRAF03012.1000EA 37.6 EUR
KRAF03012.1000 KRAF03012.2000 KRAF03012.3000
Bromophenol blue 0.1% in aqueous solution
Bromophenol blue
Formula: C₁₉H₁₀Br₄O₅S
MW: 669.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005875
CAS Number: 115-39-9
EINECS: 204-086-2