Propyzamide, Supelco®
Supplier: Merck
Warning
Synonyms: 3,5-Dichloro-N-(1,1-dimethyl-2-propynyl)benzamide,Pronamide,3,5-Dichloro-N-(2-methylbut-3-yn-2-yl)benzamide
M-Clarity™ quality level = MQ100
45645-250MG
45645-250MGEA 97.6 EUR
45645-250MG
Propyzamide, Supelco®
Propyzamide
Formula: C₁₂H₁₁Cl₂NO
MW: 256.13 g/mol
Boiling Pt: 321 °C (1013 hPa)
Melting Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00055346
CAS Number: 23950-58-5
EINECS: 245-951-4
UN: 3077
ADR: 9,III
Merck Index: 12,08058
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82c1a811dca2f8a0af5251ab0b420bf13d70bcbf&prid=web&clid=SAPDAM