- Type d'anticorps:Primaire
- Nom de l'antigène:Fibroblast growth factor 9
- Symbole de l'antigène:FGF9
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Cytométrie en flux:Yes
- Hôte:Rabbit
- Isotype:
- Réactivité:
- Western blot:Yes
- Environmentally Preferable:
- Épitope:Amino Acid Range = 31-58
- Formulaire:Liquid
- ID de gène:P31371
- Synonymes antigène:FGF9
- Modification:None
- Storage buffer:Supplied in PBS with 0,09% (W/V) sodium azide
- Molecular weight:23 kDa
- Température de stockage:Store at 4 °C for three months and –20 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
- Concentration:0,5 mg/ml
- Température de transport:4 °C
- Immunogène:This FGF9 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 31-58 amino acids from the N-terminal region of human FGF9.
- Purification:This antibody is purified through a protein A column, followed by peptide affinity purification.
- Taille:400 µl
- Cdt:400 µl
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein was isolated as a secreted factor that exhibits a growth-stimulating effect on cultured glial cells. In nervous system, this protein is produced mainly by neurons and may be important for glial cell development. Expression of the mouse homolog of this gene was found to be dependent on Sonic hedgehog (Shh) signaling. Mice lacking the homolog gene displayed a male-to-female sex reversal phenotype, which suggested a role in testicular embryogenesis.
For WB starting dilution is: 1:1000. For FACS starting dilution is: 1:10~50.
Type: Primary
Antigen: FGF9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit Ig
Reactivity: Human, Mouse