Brilliant cresyl blue zinc chloride double salt, Certistain® for microscopy, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Amino-7-(diethylamino)-2-methylphenoxazin-5-ium tetrachlorozincate (2:1)

1.01368.1000
1.01368.1000EA 2610 EUR
1.01368.1000
Brilliant cresyl blue zinc chloride double salt, Certistain® for microscopy, Sigma-Aldrich®
Brilliant Cresyl Blue
Formula: C₃₄H₄₀Cl₄N₆O₂Zn
MW: 771.93 g/mol
Melting Pt: 195…198 °C
CAS Number: 51716-96-2
EINECS: 257-358-8
IVDLogo

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR