(4-Nitrophenyl) β-D-glucopyranoside

Supplier: Roth Carl

Synonyms: p-Nitrophenyl-β-D-Glucopyranoside,(2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(4-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol

4082.1
ROTH4082.1EA 59.5 EUR
ROTH4082.1
(4-Nitrophenyl) β-D-glucopyranoside
(4-Nitrophenyl) β-D-glucopyranoside
Formula: C₁₂H₁₅NO₈
MW: 301.25 g/mol
MDL Number: MFCD00006593
CAS Number: 2492-87-7
EINECS: 219-661-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR