Sudan orange G, Supelco®

Supplier: Merck

Synonyms: Solvent Orange 14-(Phenylazo)resorcinol

M-Clarity™ quality level = MQ100

43207-25MGEA 131 EUR
43207-25MG
Sudan orange G, Supelco®
Sudan Orange G
Formula: C₁₂H₁₀N₂O₂
MW: 214.22 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 170 °C
MDL Number: MFCD00002275
CAS Number: 2051-85-6
EINECS: 218-131-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR