Sudan IV

Supplier: Roth Carl
Warning

Synonyms: Oil Red,1-((2-Methyl-4-(o-tolyldiazenyl)phenyl)diazenyl)naphthalen-2-ol,Scarlet Red,Oil Red BB,1-({2-Methyl-4-[(2-methylphenyl)diazenyl]-phenyl}diazenyl)-2-naphthol,Scarlet R,Biebrich scarlet R,Lipid Crimson,Scarlet Red Scharlach,Oil Red IV,Fat Red B,Solvent Red 24,1-(2-Methyl-4-(2-methylphenylazo)phenylazo)-2-naphthol

0327.1
ROTH0327.1EA 27.9 EUR
ROTH0327.1
Sudan IV
Sudan IV
Formula: C₂₄H₂₀N₄O
MW: 380.45 g/mol
Melting Pt: 188 °C
Storage Temperature: Ambient
MDL Number: MFCD00003893
CAS Number: 85-83-6
EINECS: 201-635-8
Merck Index: 12,08538

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR