Pyridoxine, EMPROVE® ESSENTIAL, SAFC®
Synonyms: Vitamine B6 base,Pyridoxol,(5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol
5.01260.1000
5.01260.1000EA 3200 EUR
5.01260.1000
Pyridoxine, EMPROVE® ESSENTIAL, SAFC®
Pyridoxine
Formula: C₈H₁₁NO₃
MW: 169.18 g/mol
Melting Pt: 156…157 °C
Density: 1.38 g/cm³ (20 °C)
MDL Number: MFCD00006335
CAS Number: 65-23-6
EINECS: 200-603-0