Tetrahydrothiophen-3-one

Supplier: Molekula

Synonyms: 4,5-Dihydrothiophen-3(2H)-one4,5-Dihydro-3(2H)-thiophenoneDihydrothiophen-3(2H)-one

MOLE15546870-25GEA 0 EUR
MOLE15546870-25G
Tetrahydrothiophen-3-one
Tetrahydrothiophen-3-one
Formula: C₄H₆OS
MW: 102.16 g/mol
MDL Number: MFCD00005412
CAS Number: 1003-04-9
EINECS: 213-698-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR