Imprimer
Water purification system, PURELAB® Option S
Fournisseur: ELGA LabWater
PURELAB® Option
171-0505EA 4732 EUR
171-0505 171-0504 171-0501 171-0499
Water purification system, PURELAB® Option S
Purificateurs d'eau Systèmes d'eau pure et ultra-pure
The PURELAB Option provides Type II+/II pure water on demand. Applications range from glassware washing and the feeding of ultrapure water systems to cell culture and media preparation.
  • Features ‘Reverse Osmosis feed optimised’ resin mixes to increase the ion exchange capacity of consumables and minimise running costs
  • Quick and easy sanitisation and replacement of consumables to reduce maintenance time
  • Can be wall-mounted or used on a bench
  • Can be used with wrap-around reservoirs to save space whilst optimising purity
  • Feed water: Potable supply (Tap Water)
  • Available with or without internal boost pump for flooded suction