Imprimer
Récipients pour collecte de crachats, avec bouchon clipsable, SputEm™
Fournisseur: Simport
SputEm™
SIMPC200GEA 436 EUR
SIMPC200G
Récipients pour collecte de crachats, avec bouchon clipsable, SputEm™
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement d'échantillons d'expectorations
The SputEm™ Sputum Collection System offers an ideal method to collect, carry, and process biological samples.
  • Available in a light green color, three narrow vertical windows allow the contents of the tube to be discreetly viewed
  • A wide base ensures great stability and prevents tipping
  • The large collection funnel is made in such a way that specimens fall directly into the graduated centrifuge tube and do not contaminate the outside threads
  • The centrifuge tube screw cap is kept sterile at all times under the snap cap on top of the funnel
A patient label is already affixed on top of the unit and can easily be transferred to the tube before leaving for the laboratory. The base can be used as a support during transit.
The unit features a base which incorporates a removable, sterile, 50mL graduated polypropylene conical tube that can withstand centrifugation up to 5000 rpm or 3000×g.
Certifications: For IVD use.
Conditionnement: Individually wrapped, 72 per case.