Imprimer
Gel staining and blot processing system, Processor Plus™
Fournisseur: Cytiva
80-6444-23EA 2392.45 EUR
80-6444-23 80-6445-18 80-6444-80
Gel staining and blot processing system, Processor Plus™
Systèmes d'électrophorèse Systèmes de coloration de gel et de traitement de transfert combinés
Processor Plus™ is a fluid delivery system that automates acrylamide gel staining and Western blot processing. Changing from one application to the other is achieved by a fast, simple switch of a tray. Select a programmed protocol and push start. Processor Plus™ will fill and empty the tray with the correct solution at the right time.
  • One system for gel staining and blot processing
  • Automated staining of Coomassie® blue or silver stain gels for proteins and nucleic acid analysis
  • Prepares blots for high sensitivity ECL and chromogenic detection
  • Programmable control of protocol, solution, volume, and processing time
  • Proven design for consistent, high-quality performance every time
Blot-processing capabilities of Processor Plus™ include trays for four mini (up to 9×9,5 cm) or two standard (up to 16×16 cm) sized blots at one time. Five preprogrammed and five user-defined blot protocols are incorporated for flexibility, and microprocessor precision ensures consistent results with low background from membrane to membrane.
Automated gel staining capabilities of Processor Plus™ include nine preprogrammed and four user-defined protocols for proteins and nucleic acids. A choice of small- and large-format tray sizes accommodates one to six mini-gels or one to two standard size gels for Coomassie® blue or silver staining.
The components behind the intelligent fluid handling of Processor Plus™ include a high-speed peristaltic pump for fast delivery and removal of reagents; a 10-port valve for the creation of multiple reagent protocols and segregation of hazardous waste; and a rhomboid-shaped tray with smooth and even rocking for rapid fluid mixing and coverage.
Protocol entry and editing is simple, with up to 30 steps allowed per protocol. A removable protocol key, which stores one protocol, protects critical protocols from accidental alteration.
A serial printer can be connected to the Processor Plus™ for report generation. The report provides information on each step including the volume of reagents used, the time (in min) for each step, and a validation report form, with spaces for sample ID, date and operator.