4-tert-Butyl-N-(2-fluorobenzyl)aniline, 97%

H57922.MDEA 43.1 EUR
ALFAH57922.MD ALFAH57922.03
4-tert-Butyl-N-(2-fluorobenzyl)aniline, 97%
Purity: 97%
CAS No.: 1042513-60-9
Molecular Formula: C17H20FN
Formula Weight: 257.35
MDL No.: MFCD11923956
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR