4-Bromo-3-fluoro-N-(3-fluorobenzyl)aniline, 97%

H57831.03EA 53.8 EUR
ALFAH57831.03 ALFAH57831.MD
4-Bromo-3-fluoro-N-(3-fluorobenzyl)aniline, 97%
Purity: 97%
Molecular Formula: C13H10BrF2N
Formula Weight: 298.13
MDL No.: MFCD20806792
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR