Dinonyl sulphide ≥97.0%
Leverancier: TCI EUROPE
TCIAN0360-25GEA
84.7
EUR
TCIAN0360-25G
Dinonyl sulphide ≥97.0%
Dinonyl sulphide
Formule:
C₁₈H₃₈S MW: 286,57 g/mol Kookpunt: 202°C Smeltpunt: 14,5 °C |
MDL-nummer:
MFCD00027342 CAS-nummer: 929-98-6 ADR: 9, |
Specificaties testresultaten
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...