Sodium butyrate 98-101%, Laboratory Reagent, J.T.Baker®
E186-05
Sodium butyrate 98-101%, Laboratory Reagent, J.T.Baker®
Sodium butyrate
Formule:
C₄H₇NaO₂ MW: 110,09 g/mol Smeltpunt: 244 °C Dichtheid: 1,324 g/cm³ (30 °C) |
MDL-nummer:
MFCD00002816 CAS-nummer: 156-54-7 EINECS: 205-857-6 |
Specificaties testresultaten
For Laboratory,Research,or Manufacturing Use | |
Preserve in well-closed containers. | |
Assay | 98.0 - 101.0 % |
Identification (by IR) | Passes Test |
Water (H₂O) | ≤ 1 % |
Preserve in well-closed containers. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...