2-Chloro-4-fluoropyridine ≥98.0%

Leverancier: TCI EUROPE
Gevaar

TCIAC2627-5GEA 82.5 EUR
TCIAC2627-5G
2-Chloro-4-fluoropyridine ≥98.0%
2-Chloro-4-fluoropyridine
Formule: C₅H₃ClFN
Kookpunt: 140°C
Vlampunt: 54°C
MDL-nummer: MFCD04039345
CAS-nummer: 34941-91-8
UN: 1993
ADR: 3,III
https://be.vwr.com/stibo/web/std.lang.all/96/69/28939669.jpg

Bestel nu

Specificaties testresultaten

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR