Rotary hybridisation oven, Bambino II™
Leverancier: Boekel Scientific
Boekel
731-0507EA 3790 EUR
731-0507 731-0508 731-0506
Rotary hybridisation oven, Bambino II™
Ovens Hybridisatieovens
The compact Bambino II™ hybridisation oven offers many of the same features of the larger, more expensive units.
  • Holds up to four 50 ml, six 15 ml or twelve 2,0/1,5 ml PP conical centrifuge tubes
  • Forced air convection delivers excellent uniformity
  • Stainless steel heating chamber with sealed floor seam for easy cleaning
  • Clear acrylic lid
Leveringsinformatie: The Bambino II™ is supplied complete with a standard carousel which holds four 50 ml conical tubes.