Bromocresol green 0,04% in water for pH determination

Leverancier: TCI EUROPE

TCIAB0579-500MLEA 31.9 EUR
TCIAB0579-500ML
Bromocresol green 0,04% in water for pH determination
Bromocresol green
Formule: C₂₁H₁₄Br₄O₅S
MW: 698,02 g/mol
MDL-nummer: MFCD00005874
CAS-nummer: 76-60-8
EINECS: 200-972-8
https://be.vwr.com/stibo/web/std.lang.all/25/20/28932520.jpg

Bestel nu

Specificaties testresultaten

Appearance Green to Dark green clear liquid to cloudy liquid
Transition interval (pH) (yellow)3.8 to 5.4 (blue)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR