VWR®, Bovine serum albumin (BSA)
Leverancier: VWR
VWR®
K719-50MLEA 146 EUR
K719-50ML K719-500ML K719-120ML
VWR®, Bovine serum albumin (BSA)
Eiwitten en peptiden
Concentrated 30% BSA solution, fraction V, cold alcohol isolation, in 0,85% NaCl, contains 0,1% Na-azide (as a stabiliser).