Bathocuproin 0,5 g/l in aqueous solution

Leverancier: Bernd Kraft

KRAF05846.6000EA 0 EUR
KRAF05846.6000
Bathocuproin 0,5 g/l in aqueous solution
Bathocuproin
Formule: C₂₆H₂₀N₂
MW: 360,46 g/mol
MDL-nummer: MFCD00004972
CAS-nummer: 4733-39-5
EINECS: 225-240-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR