Pilocarpine nitrate

Leverancier: Roth Carl

ROTH7523.1EA 165 EUR
ROTH7523.1
Pilocarpine nitrate
Pilocarpine nitrate
Formule: C₁₁H₁₇N₃O₅
MDL-nummer: MFCD00078497
CAS-nummer: 148-72-1
EINECS: 205-723-7

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR