Pilocarpine nitrate ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI EUROPE
|
Waarschuwing
|
TCIAP0399-5GEA
271
EUR
TCIAP0399-5G
Pilocarpine nitrate ≥98.0% (by HPLC, titration analysis)
Pilocarpine nitrate
Formule:
C₁₁H₁₇N₃O₅ |
MDL-nummer:
MFCD00078497 CAS-nummer: 148-72-1 EINECS: 205-723-7 |
Specificaties testresultaten
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +80.0 to +84.0 deg (C=2, H2O) |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...