Bathocuproin sulphonate disodium salt analytical reagent, Supelco®
Leverancier: Merck
M-Clarity™ quality level = MQ100
1.24518.0005EA
1190
EUR
1.24518.0005
1.24518.0001
Bathocuproin sulphonate disodium salt analytical reagent, Supelco®
Bathocuproin sulphonate disodium salt
Formule:
C₂₆H₁₈N₂Na₂O₆S₂ |
MDL-nummer:
MFCD00011679 CAS-nummer: 52698-84-7 EINECS: 258-111-7 |
Specificaties testresultaten
Solubility | (20 °C) slightly soluble |
Melting Point | 300 °C |
Molar Mass | 564.55 g/mol |
Bulk Density | 190 kg/m3 |
pH | 10 (10 g/l, H2O, 20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...