Bathocuproin

Leverancier: Molekula

MOLEM74294829EA 0 EUR
MOLEM74294829
Bathocuproin
Bathocuproin
Formule: C₂₆H₂₀N₂
MW: 360,46 g/mol
MDL-nummer: MFCD00004972
CAS-nummer: 4733-39-5
EINECS: 225-240-5

Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR