1-Furfurylpyrrole ≥98.0%
Leverancier: TCI EUROPE
|
Waarschuwing
|
TCIAF0700-5GEA
128
EUR
TCIAF0700-5G
1-Furfurylpyrrole ≥98.0%
1-Furfurylpyrrole
Formule:
C₉H₉NO MW: 147,18 Kookpunt: 79°C Vlampunt: 93°C Opslagtemperatuur: Koelkast |
MDL-nummer:
MFCD00010080 CAS-nummer: 1438-94-4 EINECS: 215-876-1 |
![]() |
Specificaties testresultaten
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...