5-Acetylsalicylamide ≥98.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAA1156-25GEA 62.7 EUR
TCIAA1156-25G
5-Acetylsalicylamide ≥98.0%
5-Acetylsalicylamide
Formule: C₉H₉NO₃
MW: 179,18
MDL-nummer: MFCD00049222
CAS-nummer: 40187-51-7
EINECS: 254-830-5
https://be.vwr.com/stibo/web/std.lang.all/90/01/28929001.jpg

Bestel nu

Specificaties testresultaten

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 222.0 to 226.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR