2-(3-Chlorophenoxy)propionic acid ≥98.0%

Leverancier: TCI EUROPE
Waarschuwing

TCIAC1094-25GEA 36.3 EUR
TCIAC1094-25G
2-(3-Chlorophenoxy)propionic acid ≥98.0%
2-(3-Chlorophenoxy)propionic acid
Formule: C₉H₉ClO₃
MDL-nummer: MFCD00009723
CAS-nummer: 101-10-0
EINECS: 202-915-2
https://be.vwr.com/stibo/web/std.lang.all/83/56/28938356.jpg

Bestel nu

Specificaties testresultaten

Purity (Neutralization titration) min. 98.0 %
Melting point 113.0 to 116.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR