Perfluorododecanoic acid 96%
Leverancier: Thermo Scientific
|
Gevaar
|
L14113.06EA
77.5
EUR
L14113.06
L14113.03
Perfluorododecanoic acid 96%
Perfluorododecanoic acid
Formule:
C₁₂HF₂₃O₂ MW: 614,10 g/mol Kookpunt: 245 °C Smeltpunt: 108…112 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD00198081 CAS-nummer: 307-55-1 EINECS: 206-203-2 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...