(-)-trans-2-tert-Butylcyclohexanol for synthesis, Sigma-Aldrich®

Leverancier: Merck


M-Clarity™ quality level = MQ200

8.24509.0500EA 89.5 EUR
8.24509.0500
(-)-trans-2-tert-Butylcyclohexanol for synthesis, Sigma-Aldrich®
(-)-trans-2-tert-Butylcyclohexanol
Formule: C₁₀H₂₀O
CAS-nummer: 98104-30-4

Bestel nu

Specificaties testresultaten

Molar Mass 156.27 g/mol
Hill Formula C10H20O

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR