3-(Boc-amino)-3-phenylpropionic acid 97%
Leverancier: Thermo Scientific
L19802.03EA
84.2
EUR
L19802.03
3-(Boc-amino)-3-phenylpropionic acid 97%
3-(Boc-amino)-3-phenylpropionic acid
Formule:
C₁₄H₁₉NO₄ MW: 265,31 Smeltpunt: 142…144 °C (ontbinding) |
MDL-nummer:
MFCD02090695 CAS-nummer: 14676-01-8 EINECS: 000-000-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...