4-Hydroxy-o-toluic acid ≥98%
Leverancier: Thermo Scientific
|
Gevaar
|
H60810.09EA
257
EUR
H60810.09
4-Hydroxy-o-toluic acid ≥98%
4-Hydroxy-o-toluic acid
Formule:
C₈H₈O₃ MW: 152,15 g/mol Smeltpunt: 177…181 °C Opslagtemperatuur: Kamertemperatuur |
MDL-nummer:
MFCD02182261 CAS-nummer: 578-39-2 EINECS: 209-422-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...