5-Cyano-2-fluorobenzeneboronic acid pinacol ester 96%

Synoniemen: 5-Cyano-2-fluorophenylboronic acid pinacol ester

H62739.03EA 419 EUR
H62739.03
5-Cyano-2-fluorobenzeneboronic acid pinacol ester 96%
5-Cyano-2-fluorobenzeneboronic acid pinacol ester
Formule: C₁₃H₁₅BFNO₂
MW: 247,07 g/mol
Opslagtemperatuur: Kamertemperatuur
MDL-nummer: MFCD07437899
CAS-nummer: 863868-29-5

Bestel nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR