Pipette containers, SPECICON
Leverancier: Schuett-biotec
SPECICON
612-1865EA 128 EUR
612-1865 612-1854 612-1866 612-1855
Pipette containers, SPECICON
Dozen Pipetsterilisatiecontainers
Containers are made of thick walled aluminium, anodised in gold colour.
  • With sterilisation opening and bayonet closures
  • Can be hermetically sealed
They can be used for the sterilisation and storage of pipettes.