Single wafer tray, H 20 series, 1,2"
Leverancier: Entegris
ENTEH20-250-1415EA 0 EUR
ENTEH20-250-1415
Single wafer tray, H 20 series, 1,2"
Wafer handling trays to safely transport and protect silicon wafers.
  • For internal handling of single wafers
  • Conical pocket holds one wafer facedown contacting only the wafer’s edges
  • Tweezer slots on tray bottom ease wafer removal
  • Stackable for efficient use of storage space, either tray on tray or cover on tray