Anti-FRS2 Mouse Polyclonal Antibody

Leverancier: Abnova
ABNOH00010818-A02EA 258 EUR
ABNOH00010818-A02
Anti-FRS2 Mouse Polyclonal Antibody
Antilichamen
Type: Primary
Antigen: FRS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR