Anti-GSK3A Rabbit Polyclonal Antibody

Leverancier: Biorbyt
BIRBORB6121-100EA 305 EUR
BIRBORB6121-100
Anti-GSK3A Rabbit Polyclonal Antibody
Antilichamen
Western Blot: 1:500-1:1000 Immunochemistry: 1:50-1:100 Immunofluorescence: 1:100-1:200

Type: Primary
Antigen: GSK3A (glycogen synthase kinase 3 alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR