Anti-HDAC7 Rabbit Polyclonal Antibody
Leverancier: Biorbyt
BIRBORB6137-200EA 0 EUR
BIRBORB6137-200 BIRBORB6137-100
Anti-HDAC7 Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: HDAC7 (histone deacetylase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu