Anti-ITK Rabbit Polyclonal Antibody

Leverancier: Biorbyt
BIRBORB6207-100EA 0 EUR
BIRBORB6207-100 BIRBORB6207-200
Anti-ITK Rabbit Polyclonal Antibody
Antilichamen
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: ITK (IL2-inducible T-cell kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestel nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR